missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ RapidCal Immuno Decalcifier

RapidCal Immuno is designed to preserve maximum immunogenicity while accomplishing rapid decalcification.

$95.70 - $287.63

Specifications

Color Clear
Product Type Decalcifying Solution
Solution Type Decalcifier
For Use With (Application) Bone Decalcification
Form Liquid
View More Specs

Alert:

Harmful if swallowed, in contact with skin, or inhaled.


Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
STL6086
View Documents
StatLab™
6086
1 pt./Bottle
Each for $95.70
Only null left
 
STL6089
View Documents
StatLab™
6089
1 gal./Bottle, 4 Bottles/Cs.
Each for $287.63
Only null left
 
Description

Description

  • Formulated to decalcify bone rapidly
  • Performs gently enough to be used as a routine decalcifying solution for bone marrow biopsies
Specifications

Specifications

Clear
Decalcifier
Liquid
Decalcifying Solution
Bone Decalcification
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'