missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab™ RapidCal Immuno Decalcifier
Description
- Formulated to decalcify bone rapidly
- Performs gently enough to be used as a routine decalcifying solution for bone marrow biopsies
Specifications
Specifications
| Color | Clear |
| Product Type | Decalcifying Solution |
| Quantity | 1 pt./Bottle |
| Solution Type | Decalcifier |
| For Use With (Application) | Bone Decalcification |
| Form | Liquid |