missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapID™ Nitrate A Reagent

Catalog No. R8309003
Change view
Click to view available options
Quantity:
15 mL/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R8309003 15 mL/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R8309003 Supplier Thermo Scientific™ Supplier No. R8309003

Complete the Thermo Scientific™ RapID™ System Nitrite test using the ready-to-use Thermo Scientific RapID Nitrate A Reagent.

Complete the Thermo Scientific™ RapID™ System Nitrite test using the Thermo Scientific RapID Nitrate A Reagent.

This product is for use with RapID CB PLUS System (R8311008), RapID NF PLUS System (R8311005), RapID ONE System (R8309002), RapID NH System (R8311001), RapID STAPH PLUS System (R8309003) and RapID NH System (R8311001).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Content And Storage 2°C to 8°C
Description RapID Nitrate A Reagent
CE Marker Yes
Format Vial
For Use With (Equipment) RapID CB PLUS System (R8311008), RapID NF PLUS System (R8311005), RapID ONE System (R8309002), RapID NH System (R8311001), RapID STAPH PLUS System (R8309003, RapID NH System (R8311001)
Packaging Type 1 Box
Quantity 15 mL/Vial