missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapID™ 2 mL Inoculation Fluid

Catalog No. R8325106
Change view
Click to view available options
Quantity:
20 Tubes/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R8325106 20 Tubes/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R8325106 Supplier Thermo Scientific™ Supplier No. R8325106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Save time preparing diluents with this ready-to-use inoculation fluid for RapID systems.

Save time preparing diluents for Thermo Scientific™ RapID™ System using Thermo Scientific RapID 2 mL Inoculation Fluid.

This product is for use with RapID ONE System (R8311006), RapID YEAST PLUS System (R8311007), RapID STR System (R8311003), RapID CB PLUS System (R8311008) and RapID STAPH PLUS System (R8311009).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Content And Storage 20°C to 25°C
Description RapID 2mL Inoculation Fluid
For Use With (Equipment) RapID ONE System (R8311006), RapID YEAST PLUS System (R8311007), RapID STR System (R8311003), RapID CB PLUS System (R8311008), RapID STAPH PLUS System (R8311009)
Packaging Type 1 Box
Quantity 20 Tubes/Pk.