missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapID™ 1mL Inoculation Fluid

Catalog No. R8325102
Change view
Click to view available options
Quantity:
20 Tubes/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R8325102 20 Tubes/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R8325102 Supplier Thermo Scientific™ Supplier No. R8325102

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Save time preparing diluents with this ready-to-use inoculation fluid for RapID systems.

Save time preparing diluents for Thermo Scientific™ RapID™ System using Thermo Scientific RapID 1 mL Inoculation Fluid.

This product is for use with RapID NH System (R8311001), RapID ANA II System (R8311002), RapID STR System (R8311003), RapID SS/u System (R8311004) and RapID NF PLUS System (R8311005).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Content And Storage 20°C to 25°C
Description RapID 1mL Inoculation Fluid
For Use With (Equipment) RapID NH System (R8311001), RapID ANA II System (R8311002), RapID STR System (R8311003), RapID SS/u System (R8311004), RapID NF PLUS System (R8311005)
Packaging Type 1 Box
Quantity 20 Tubes/Pk.