missing translation for 'onlineSavingsMsg'
Learn More

Rapamycin, 99.0%, MP Biomedicals™

Catalog No. ICN15934680
Change view
Click to view available options
Quantity:
100 μg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15934680 100 μg
ICN15934691 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15934680 Supplier MP Biomedicals Inc Supplier No. 0215934680

Chemical Identifiers

CAS 53123-88-9
Molecular Formula C51H79NO13
Molecular Weight (g/mol) 914.187
InChI Key QFJCIRLUMZQUOT-HPLJOQBZSA-N
Synonym A422989, NSC226080
PubChem CID 5284616
ChEBI CHEBI:9168
SMILES CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)O)C)C)O)OC)C)C)C)OC

Specifications

Type Rapamycin
Physical Form Powder
Color White
Quantity 100 μg
Assay Percent Range 0.99
Formula Weight 914.2
Chemical Name or Material Rapamycin