missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Rap1 Activation Assay Kit

Catalog No. 17321
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17321 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17321 Supplier MilliporeSigma™ Supplier No. 17321

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • 100X GTPγS, 10mM
  • 100X GDP, 100mM
  • Rap1 Activation Lysis Buffer, 2X
  • Anti-Rap1
  • Rap1 Assay Reagent (Ral GDS-RBD, agarose)

Non-radioactive Kit that uses Ral GDS RBD, agarose to precipitate Rap1-GTP from cell lysates and detection by a Rap1 specific antibody

  • Non-radioactive Kit that uses Ral GDS RBD, agarose to precipitate Rap1-GTP from cell lysates and detection by a Rap1 specific antibody
  • Applications: Affinity Binding Assay

Specifications

Detection Method Non-radioactive Method
For Use With (Application) Affinity Binding Assay
Quantity 1 Kit
Type Activation Assay Kit
Cell Line HEK 293