missing translation for 'onlineSavingsMsg'
Learn More

Ranitidine Hydrochloride 98.0+%, TCI America™

Catalog No. R00735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00735G 5 g
R007325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R00735G Supplier TCI America Supplier No. R00735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66357-59-3
Molecular Formula C13H23ClN4O3S
Molecular Weight (g/mol) 350.862
MDL Number MFCD00069339
InChI Key GGWBHVILAJZWKJ-CHHCPSLASA-N
Synonym ranitidine hydrochloride, noctone, n-2-5-dimethylamino methyl-2-furanyl methyl thio ethyl-n'-methyl-2-nitro-1,1-ethanediamine hydrochloride, c13h22n4o3s.hcl, opera_id_624, melfax hydrochloride, zantac hydrochloride, zintac hydrochloride, azantac hydrochloride, raniben hydrochloride
PubChem CID 6603542
IUPAC Name (Z)-1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride
SMILES CNC(=C[N+](=O)[O-])NCCSCC1=CC=C(O1)CN(C)C.Cl

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 5 g
Formula Weight 350.86
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ranitidine Hydrochloride
RTECSNumber KM6557000
TSCA No
Recommended Storage Refrigerator