missing translation for 'onlineSavingsMsg'
Learn More

Ranitidine hydrochloride, 99%

Catalog No. p-7026316
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2226006 5 g
AAB2226014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2226006 Supplier Thermo Scientific Chemicals Supplier No. B2226006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 66357-59-3 | C13H23ClN4O3S | 350.862 g/mol

An H-2 histamine receptor antagonistRanitidine hydrochloride is mainly used as an H2-receptor antagonist and helps in the treatment of gastrointestinal lesions because of excessive gastric acid secretion. It also serves as an antiulcer drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An H-2 histamine receptor antagonistRanitidine hydrochloride is mainly used as an H2-receptor antagonist and helps in the treatment of gastrointestinal lesions because of excessive gastric acid secretion. It also serves as an antiulcer drug.

Solubility
Soluble in water, 2-hydroxypropyl-beta-cyclodextrin, acetic acid, methanol, ethanol and dimethyl sulfoxide. Insoluble in chloroform.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 66357-59-3
Molecular Formula C13H23ClN4O3S
Molecular Weight (g/mol) 350.862
MDL Number MFCD00069339
InChI Key GGWBHVILAJZWKJ-CHHCPSLASA-N
Synonym ranitidine hydrochloride, noctone, n-2-5-dimethylamino methyl-2-furanyl methyl thio ethyl-n'-methyl-2-nitro-1,1-ethanediamine hydrochloride, c13h22n4o3s.hcl, opera_id_624, melfax hydrochloride, zantac hydrochloride, zintac hydrochloride, azantac hydrochloride, raniben hydrochloride
PubChem CID 6603542
IUPAC Name (Z)-1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride
SMILES CNC(=C[N+](=O)[O-])NCCSCC1=CC=C(O1)CN(C)C.Cl

Specifications

Melting Point ∽134°C (decomposition)
Assay Percent Range 99%
Quantity 5 g
Merck Index 14,8110
Solubility Information Soluble in water,2-hydroxypropyl-beta-cyclodextrin,acetic acid,methanol,ethanol and dimethyl sulfoxide. Insoluble in chloroform.
Formula Weight 350.86
Percent Purity 99%
Chemical Name or Material Ranitidine hydrochloride
EINECSNumber 266-333-0
TSCA No
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only