missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ramipril, 98%

Catalog No. AAH3692603
Change view
Click to view available options
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AAH3692603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3692603 Supplier Thermo Scientific™ Supplier No. H36926MD
Only null left

Chemical Identifiers

CAS 87333-19-5
Molecular Formula C23H32N2O5
Molecular Weight (g/mol) 416.52
MDL Number MFCD00865775
InChI Key HDACQVRGBOVJII-JBDAPHQKSA-N
IUPAC Name (2S,3aS,6aS)-1-[(2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]-octahydrocyclopenta[b]pyrrole-2-carboxylic acid
SMILES CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1[C@H]2CCC[C@H]2C[C@H]1C(O)=O

Specifications

Quantity 1 g
Formula Weight 416.52
Percent Purity 98%
Physical Form Solid
ShelfLife 5 Years
EINECSNumber 642-904-3
RTECSNumber GY5879600