Learn More
Thermo Scientific™ MarqMetrix All-In-One Process Raman Analyzer
The Thermo Scientific™ MarqMetrix™ All-In-One Process Raman Analyzer is a compact and portable Raman spectrometer all-in-one system with patented technology, purpose-built for rapid deployment, ease-of-use, and flexible scalability in markets where time-to-results is key.
Supplier: Thermo Scientific™ 912A1261
Description
With the MarqMetrix Process Raman Analzyer, businesses can conduct real-time, non-destructive, and immediate analysis without sample preparation.
The MarqMetrix All-In-One analyzer makes precise and accurate measurements simple to execute and understand. Our one-button hardware and easy-to-use software will have you taking highly accurate Raman measurements in less than 15 minutes out of the box.
The MarqMetrix All-In-One System is easily integrated into your existing process and eliminates the need for costly technical expertise.
Application versatility
- Bioprocessing PAT (Process Analytical Technology)
- Oil & Gas production
Use Raman inline for process monitoring
With a small footprint and lower cost, the MarqMetrix All-In-One Process Raman Analyzer makes process monitoring possible in-line and enables you to make optimization decisions faster.
Pack your MarqMetrix All-In-One System in a protective case and take it to the point of need. Factory calibration ensures continuous and precise analysis out of the box.
Service Plans
Thermo Fisher Scientific offers Service Plans designed to maximize productivity and optimize performance throughout the lifecycle of your MarqMetrix All-In-One system. Plans include exclusive service advantages such as:
- Unlimited technical support
- Depot repair services
- Maintaining system compliance
- Consulting services
Software
Maximize the power of the MarqMetrix All-In-One Process Raman Analyzer with Thermo Scientific Lykos PAT Software - a remote control and monitoring software designed to simplify the complexities of chemometrics with features including:
- Real-time display, storage, and export of bioprocess monitoring results
- Remotely control and monitor the MarqMetrix All-In-One Process Raman Analyzer from outside of the lab
- Simplified use and intuitive workflow for non-Ramen spectroscoptists
- Control multiple MarqMetrix All-In-One analyzers from a central location
Available add-ons: cGMP and 21 CFR part 11 compliance features for regulated environments
Chemical Identifiers
| Process Raman Analyzer | |
| Input voltage: 90-264 VAC, 50-60 Hz, Operating voltage: 12VDC, 2A | |
| 6.5 cm-1 average across spectral range | |
| 100 to 3250 cm-1 | |
| 25 cm | |
| Thermally stabilized CCD (2 stage TEC) | |
| Laser Certification: Class 3B laser deviceReported to the FDA | |
| 1RJ-45 Ethernet port 3 USB 3.0, 1 Display port 1.1 (VGA, DVI or HDMI with adaptor) |
| 25 cm | |
| 7.7 cm | |
| Software: MarqMetrix Raman suiteController: Integrated PC (Windows 10 OS) | |
| 785 nm, Excitation power: 10 to 450 mW | |
| 1 keyed Laser interlock switch, Electronic laser trigger | |
| FC/PC (Both excitation and collection) | |
| ≤16.5 counts at 10 sec. integration |
Specifications
| Process Raman Analyzer | |
| Input voltage: 90-264 VAC, 50-60 Hz, Operating voltage: 12VDC, 2A | |
| 6.5 cm-1 average across spectral range | |
| 100 to 3250 cm-1 | |
| 25 cm | |
| Thermally stabilized CCD (2 stage TEC) | |
| Process Ball Probe | |
| ≤16.5 counts at 10 sec. integration |
| 25 cm | |
| 7.7 cm | |
| Software: MarqMetrix Raman suiteController: Integrated PC (Windows 10 OS) | |
| 785 nm, Excitation power: 10 to 450 mW | |
| 1 keyed Laser interlock switch, Electronic laser trigger | |
| FC/PC (Both excitation and collection) | |
| Laser Certification: Class 3B laser deviceReported to the FDA | |
| 1RJ-45 Ethernet port 3 USB 3.0, 1 Display port 1.1 (VGA, DVI or HDMI with adaptor) |