missing translation for 'onlineSavingsMsg'
Learn More

RAM Scientific Safe-T-Fill™ Plastic Hematocrit Capillaries

Catalog No. 1491581
Change view
Click to view available options
Additive:
None
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Additive
1491581 None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1491581 Supplier RAM Scientific Supplier No. 06 0508

100% plastic—no glass to break

Blood samples are drawn into the SAFE-T-FILL™ Plastic Microhematocrit Capillary Tubes through the same capillary action as glass without the risk of breakage.

  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Replaces glass Natelson, Caraway, or mylar-coated glass microhematocrit capillary tubes
  • Blood samples are drawn into the capillary tube through the same capillary action as glass without the risk of breakage
  • Microhematocrit capillary tubes are available untreated or heparinized
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • Color-coded tip red/blue marking to designate whether additive is present in the capillary tube
  • Packaging labels are marked with Lot Number and Expiration Date

Recommended Storage

Store at room temperature. Keep dry. Keep away from sunlight.

Specifications

Color Clear with Red Mark
Product Type Microhematocrit Untreated Capillary Tube
Additive None
Material Plastic
Capacity (Metric) 50 μL
Closure Included No
For Use With (Application) Intended for microhematocrit determinations (volume of red blood cells/PCV).
Length (Metric) 7.5 cm
Length (English) 2.95 in.