missing translation for 'onlineSavingsMsg'
Learn More

RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: EDTA

Catalog No. 1491586
Change view
Click to view available options
Capacity (Metric):
200 μL
300 μL
Additive:
K2 EDTA
Product Type:
Hematology/Whole Blood Tube
Self-sealing Whole Blood Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Additive Product Type
1491586 200 μL K2 EDTA Self-sealing Whole Blood Tube
1491553 300 μL K2 EDTA Hematology/Whole Blood Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1491586 Supplier RAM Scientific Supplier No. 07 7056

No scooping needed, allowing for high-quality, tissue-free samples

Blood samples are drawn into the SAFE-T-FILL™ Plastic Micro Capillary Blood Collection Tubes through capillary action, reducing hemolysis,

  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No "scooping" or "milking" required
  • Capillary straw and microtube are coated with anticoagulant Dipotassium (K2) EDTA (Purple cap) to prevent clotting
  • Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
  • Lot Number, Expiration Date, and fill lines are printed directly on the microtube
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • For whole blood hematology determinations
  • Small sample sizes, great for pediatrics and neonates
  • Available with self-sealing rubber cap (Not made with natural rubber latex)
  • Available in a variety of sizes and draw volumes

Recommended Storage

Store at room temperature. Keep dry. Keep away from sunlight.

Specifications

Color Clear
Product Type Self-sealing Whole Blood Tube
Additive K2 EDTA
Material Plastic
Sterility Non-sterile
Closure Type Self-sealing Pierceable Cap
Capacity (Metric) 200 μL
Closure Included Yes
Closure Color Purple
Quantity 50 Tubes/Bag, 10 Bags/Cs.
For Use With (Application) Intended for hematology (haematology) determinations that require whole blood samples. Self-sealing cap ensures automatic closure, facilitating transport and prevents contamination.
Show More Show Less