Learn More
RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: EDTA
No scooping needed, allowing for high-quality, tissue-free samples
Supplier:  RAM Scientific 077058
 View more versions of this product
Catalog No. 14-915-54
Only null left
Description
Description
Blood samples are drawn into the SAFE-T-FILL™ Plastic Micro Capillary Blood Collection Tubes through capillary action, reducing hemolysis,
  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No "scooping" or "milking" required
  • Capillary straw and microtube are coated with anticoagulant Dipotassium (K2) EDTA (Purple cap) to prevent clotting
  • Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
  • Lot Number, Expiration Date, and fill lines are printed directly on the microtube
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • For whole blood hematology determinations
  • Small sample sizes, great for pediatrics and neonates
  • Available with self-sealing rubber cap (Not made with natural rubber latex)
  • Available in a variety of sizes and draw volumes
Recommended Storage
Store at room temperature. Keep dry. Keep away from sunlight.
Specifications
Specifications
Clear
K2 EDTA
Non-sterile
200 μL
Purple
Intended for hematology (haematology) determinations that require whole blood samples. Flat bottom tube facilitates testing and transport.
Flat-bottom Whole Blood Tube
Plastic
Standard Pierceable Cap
Yes
50 Tubes/Bag, 10 Bags/Cs.
Product Suggestions
Product Suggestions