missing translation for 'onlineSavingsMsg'
Learn More

Raltegravir potassium salt

Catalog No. p-7112905
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462162500 250 mg
AC462160010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462162500 Supplier Thermo Scientific Chemicals Supplier No. 462162500
Only null left

CAS: 871038-72-1 | C20H20FKN6O5 | 482.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 871038-72-1
Molecular Formula C20H20FKN6O5
Molecular Weight (g/mol) 482.51
InChI Key IFUKBHBISRAZTF-UHFFFAOYSA-M
Synonym raltegravir potassium, raltegravir potassium salt, raltegravirpotassiumsalt, raltegravir mk-0518, potassium 4-4-fluorobenzyl carbamoyl-1-methyl-2-2-5-methyl-1,3,4-oxadiazole-2-carboxamido propan-2-yl-6-oxo-1,6-dihydropyrimidin-5-olate, isentress tn, raltegravir potassium usan:jan, pubchem22484, n-1-4-4-fluorophenyl methylcarbamoyl-5-hydroxy-1-methyl-6-oxo-pyrimidin-2-yl-1-methyl-ethyl-5-methyl-1,3,4-oxadiazole-2-carboxamide
PubChem CID 23668479
IUPAC Name potassium;4-[(4-fluorophenyl)methylcarbamoyl]-1-methyl-2-[2-[(5-methyl-1,3,4-oxadiazole-2-carbonyl)amino]propan-2-yl]-6-oxopyrimidin-5-olate
SMILES CC1=NN=C(O1)C(=O)NC(C)(C)C2=NC(=C(C(=O)N2C)[O-])C(=O)NCC3=CC=C(C=C3)F.[K+]

Specifications

Melting Point 155.0°C to 157.0°C
Quantity 250 mg
Formula Weight 482.51
Chemical Name or Material Raltegravir potassium salt
Hazard Category Serious eye damage/eye irritation (category 1), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Reproductive toxicity (category 2), Skin corrosion/irritation (category 2)
Hazard Statement Causes serious eye damage, Harmful if swallowed, May cause respiratory irritation, Suspected of damaging fertility or the unborn child, Causes skin irritation

RUO – Research Use Only