missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Rainbow™ Molecular Weight Markers
Description
Enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels. The ready-to-load markers provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination.
- High intensity, prestained multicolored bands allow distinction of closely related proteins
- Full-Range Rainbow Markers (Mr 12,000 to 225,000) use ten separate proteins with six different colors
- High-Range Rainbow Markers (Mr 12,000 to 225,000) use eight separate proteins with six different colors
- Low-Range Rainbow Markers (Mr 3500 to 38,000) use seven separate proteins with five different colors
- For use on SDS-PAGE
- The bright, distinctive colors of the markers allow easier confirmation of transfer to blotting membranes and orientation.
- Markers are a mixture of individually colored proteins that are combined to produce bands of equal color intensity. Approximate protein concentration is 1 to 2 mg/mL, depending on the product
- Each 250μL pack size is sufficient for use with 50 minigels (10 × 8cm) or 25 large gels (20 × 20cm)
Specifications
Specifications
| Description | Rainbow molecular weight markers enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels. |
| Color | Multi |
| Concentration | 1.5 mg/mL |
| Form | Liquid |
| Formulation | Supplied ready to use in 305% Glycerol and sample Buffer containing Mercaptoethanesulphonic Acid (MESNA) as reducing agent |
| For Use With (Application) | For industrial applications: Analytical reagent, research Use eight separate proteins with six different colors |
| Molecular Weight (g/mol) | Mr 12 000 |
| Quantity | 250 μL |
| Range | 12000 to 225000Mr |
| Storage Requirements | -15 to -30°C |
| Show More |