missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RAF1 (Y340D/Y341D) (Human) Recombinant Protein
Description
Human RAF1 (NP_002871.1, 306 a.a. - 648 a.a.) Y340D/Y341D mutant partial recombinant protein with GST-tag at N-terminal using baculovirus expression system.
Specifications
Specifications
| For Use With (Application) | Functional Study |
| Gene ID (Entrez) | 5894 |
| Name | v-raf-1 murine leukemia viral oncogene homolog 1 |
| Quality Control Testing | The purity was assessed by SDS-PAGE/CBB staining. |
| Quantity | 5 μg |
| Storage Requirements | Stored at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | CRAF, NS5, Raf-1, c-Raf |