missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph™ Radleys AVA Pad Reactor Controller
Description
Combining the proven technology of the Mya 4 Control Pad, a Data Hub, and AVA Software into a compact all-in-one controller.
- 10 in. Touchscreen display with pinch zoom
- Integrated connections (4 x RS232, 2 x Lemo, 2 x USB)
- HDMI Out to connect larger external display
- Use the software flip screen feature to mount the Control
- Pad either way up to suit your requirements
- Designed to control either single or dual reactors
- User management with individual password protection
- Network share function
- No License requirement - Any updates via 'Anytime' Upgrade
Specifications
Specifications
| For Use With (Equipment) | Lab Reactors |
| Includes | Compact 10 in. Touchscreen |
| Vessel Type | Not Applicable |
| Product Type | Reactor Controller |
| Voltage | 115 V |