Learn More
Alkali Scientific Radiant™ Probe 1-Step Lo-ROX qPCR Kit, 300 Reactions, (3 X 1mL Probe 1-Step Lo-ROX 2X qPCR Mix, 3 x 0.2mL 20X RTase with RNase Inhibitor)
Catalog No. 502389038
Encompass
Alkali Scientific offers Radiant™ Probe 1-Step Lo-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for best-in- class quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The proprietary buffer system allows for highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low- copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Probe 1-Step Lo-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection.
Catalog No. 502389038 Supplier Alkali Scientific Supplier No. QL4003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alkali Scientific offers Radiant™ Probe 1-Step Lo-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for best-in- class quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The proprietary buffer system allows for highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low- copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Probe 1-Step Lo-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection.