Learn More
Alkali Scientific Radiant™ HRM Mix, High resolution melt - SNP detection, 500 Reactions, (5 x 1mL)
Catalog No. 503028350
Encompass
Radiant™HRM Mix produces ultra-sensitive melt profiles allowing accurate discrimination subtle sequence variations including class I to class IV mutations and CpG methylation differences. Fast and accurate detection of subtle sequence differences Ultra-sensitive fluorescence profiles for clear characterisation of samples Third generation saturating dye SyGreen 2 Antibody-mediated hot start technology for improved sensitivity Compatible with all real-time instruments with HRM capability
Catalog No. 503028350 Supplier Alkali Scientific Supplier No. QHRM5
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Radiant™HRM Mix produces ultra-sensitive melt profiles allowing accurate discrimination subtle sequence variations including class I to class IV mutations and CpG methylation differences. Fast and accurate detection of subtle sequence differences Ultra-sensitive fluorescence profiles for clear characterisation of samples Third generation saturating dye SyGreen 2 Antibody-mediated hot start technology for improved sensitivity Compatible with all real-time instruments with HRM capability