Learn More
Alkali Scientific Radiant Hot Start Taq Polymera
Catalog No. 503026571
Encompass
Radiant Hot Start Taq DNA Polymerase is a highly purified high performance Hot-Start DNA Polymerase optimized for the sensitive DNA amplification of a wide range of DNA templates including complex mammalian genomic DNA and crude samples The Hot-Start technology is based on a new-generation PCR buffer formulation in conjunction with a proprietary antibody-mediated chemistry Radiant Hot-Start Taq Polymerase exhibits 5-3 DNA polymerase activity with an error-rate of wild-type Taq (2 0 x 10-5) The polymerase is ideal for genotyping colony PCR multiplexing low-copy assays and the amplification of challenging targets susceptible to mispriming RadiantTM Hot Start Taq DNA Polymerase is engineered for robust superior PCR and is supplied with a highly optimized new-generation 5x buffer system which provides exceptional sensitivity and ease of use New-generation 10X PCR buffer MgCl2 and enhancers for maximum PCR efficiency and reaction speed Robust PCR performance across a wide range of DN
Catalog No. 503026571 Supplier Alkali Scientific Supplier No. C321
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Radiant Hot Start Taq DNA Polymerase is a highly purified high performance Hot-Start DNA Polymerase optimized for the sensitive DNA amplification of a wide range of DNA templates including complex mammalian genomic DNA and crude samples The Hot-Start technology is based on a new-generation PCR buffer formulation in conjunction with a proprietary antibody-mediated chemistry Radiant Hot-Start Taq Polymerase exhibits 5-3 DNA polymerase activity with an error-rate of wild-type Taq (2 0 x 10-5) The polymerase is ideal for genotyping colony PCR multiplexing low-copy assays and the amplification of challenging targets susceptible to mispriming RadiantTM Hot Start Taq DNA Polymerase is engineered for robust superior PCR and is supplied with a highly optimized new-generation 5x buffer system which provides exceptional sensitivity and ease of use New-generation 10X PCR buffer MgCl2 and enhancers for maximum PCR efficiency and reaction speed Robust PCR performance across a wide range of DN