Learn More
Alkali Scientific Radiant HiFi Polymerase Pro 2X
Catalog No. 503027035
Encompass
RadiantHiFi Polymerase Pro2X RED Master Mix is a high-performance PCR solution designed for precision and efficiency This ready-to-use mastermix contains HiFi polymerase optimized buffer dNTPs and advanced PCR enhancers ensuring robust amplification with exceptional fidelity The red dye simplifies gel loading and visual tracking Improved DNA binding and increased processivity result in shorter extension times higher yields and the ability to amplify longer and more difficult targets High temperature cycling up to 100 C denaturation to better separate GC-rich templates Increased PCR success rates with complex genomic templates (17 5 kb and over) High yields under standard and fast PCR conditions (10-30 s/kb) Efficient and specific amplification from challenging templates including GC and AT-rich sequences 100x higher fidelity than Taq DNA polymerase Generates blunt-end PCR products Available as a convenient 2x ready mix with the option of a red dye for direct gel loading
Catalog No. 503027035 Supplier Alkali Scientific Supplier No. HPF810
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
RadiantHiFi Polymerase Pro2X RED Master Mix is a high-performance PCR solution designed for precision and efficiency This ready-to-use mastermix contains HiFi polymerase optimized buffer dNTPs and advanced PCR enhancers ensuring robust amplification with exceptional fidelity The red dye simplifies gel loading and visual tracking Improved DNA binding and increased processivity result in shorter extension times higher yields and the ability to amplify longer and more difficult targets High temperature cycling up to 100 C denaturation to better separate GC-rich templates Increased PCR success rates with complex genomic templates (17 5 kb and over) High yields under standard and fast PCR conditions (10-30 s/kb) Efficient and specific amplification from challenging templates including GC and AT-rich sequences 100x higher fidelity than Taq DNA polymerase Generates blunt-end PCR products Available as a convenient 2x ready mix with the option of a red dye for direct gel loading