Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
Catalog No. 11679084
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Holds:
10 Canisters (for cord blood, Model 4000610)
161 1.2 mL/2 mL Ampules
161 4 mL/5 mL Ampules
3 x 4/5 mL Cryo Racks (Large)
3 x 4/5 mL Cryo Racks (Small)
40 0.25/0.5 mL Straws
5 x 1.2/2 mL Cryo Racks (Large)
5 x 1.2/2 mL Cryo Racks (Small)
76 1.2 mL/2 mL Ampules
76 4 mL/5 mL Ampules
Adjustable, 10-position
Type:
Canister Rack
Straw Rack
Vial Rack (Large)
Vial Rack (Small)
Vial Rack Holder
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Type
11679084 Adjustable, 10-position Canister Rack
15467421 40 0.25/0.5 mL Straws Straw Rack
TF4000340 10 Canisters (for cord blood, Model 4000610) Canister Rack
15467461 76 1.2 mL/2 mL Ampules Vial Rack (Small)
15467462 5 x 1.2/2 mL Cryo Racks (Small) Vial Rack Holder
15467463 161 1.2 mL/2 mL Ampules Vial Rack (Large)
15467464 5 x 1.2/2 mL Cryo Racks (Large) Vial Rack Holder
13262837 76 4 mL/5 mL Ampules Vial Rack (Small)
13262838 3 x 4/5 mL Cryo Racks (Small) Vial Rack Holder
13262839 161 4 mL/5 mL Ampules Vial Rack (Large)
13262840 3 x 4/5 mL Cryo Racks (Large) Vial Rack Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 missing translation for 'options'
Catalog No. 11679084 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 185089
Get Thermo Scientific Accessories with Purchase
View all promotions
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
  • Warranty varies by country, please contact us for details
Specifications
Description 10 position, adjustable
For Use With (Equipment) All CryoMed controlled-rate freezers
Holds Adjustable, 10-position
Type Canister Rack