Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
$702.15 - $1,199.33
| Catalog Number | Mfr. No. | Type | Holds | For Use With (Equipment) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Holds | For Use With (Equipment) | Price | Quantity & Availability | |||
|
13874770
|
Thermo Scientific™
1600002 |
Rack | Depends on insert purchased | A5B,A10B A24B,S49,S19T,S14P,S21P |
Each for $1,100.97
|
|
|||
|
1325580
|
Thermo Scientific™
1600003 |
Rack Insert | 100-10 mm Tubes | A5B,A10B A24B,S49,S19T,S14P,S21P |
Each for $834.45
|
|
|||
|
1325560
|
Thermo Scientific™
1600004 |
Rack Insert | 60-16 mm Tubes | A5B,A10B A24B,S49,S19T,S14P,S21P |
Each for $834.45
|
|
|||
|
1325561
|
Thermo Scientific™
1600005 |
Rack and Rack Insert | 25-10 mm Tubes | A5B,A10B A24B,S49,S19T,S14P,S21P |
Each for $923.30
|
|
|||
|
1325562
|
Thermo Scientific™
1600006 |
Rack Insert | Dishes, plates, bags, pouches | A5B,A10B A24B,S49,S19T,S14P,S21P |
Each for $714.76
|
|
|||
|
1325563
|
Thermo Scientific™
1600079 |
Rack | Depends on insert purchased | A25B, A40, S21, S30 |
Each for $1,100.97
|
|
|||
|
1325565
|
Thermo Scientific™
1600081 |
Rack Insert | 32-16 mm Tubes | A25B, A40, S21, S30 |
Each for $1,016.55
|
|
|||
|
13255651
|
Thermo Scientific™
1600082 |
Rack Insert | 13-25 mm Tubes | A25B, A40, S21, S30 |
Each for $923.30
|
|
|||
|
13255652
|
Thermo Scientific™
1600083 |
Rack Insert | Dishes, plates, bags, pouches | A25B, A40, S21, S30 |
Each for $735.28
|
|
|||
|
13255653
|
Thermo Scientific™
1600026 |
Rack | Depends on insert purchased | S13, S12T, S15 |
Each for $1,199.33
|
|
|||
|
13255654
|
Thermo Scientific™
1600084 |
Rack Insert | 39-10 mm Tubes | S13, S12T, S15 |
Each for $1,014.06
|
|
|||
|
13255655
|
Thermo Scientific™
1600085 |
Rack Insert | 20-16 mm Tubes | S13, S12T, S15 |
Each for $923.30
|
|
|||
|
13872270
|
Thermo Scientific™
1600087 |
Rack Insert | Dishes, plates, bags, pouches | S13, S12T, S15 |
Each for $702.15
|
|
|||
Description
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Specifications
| Stainless Steel |