Learn More
Thermo Scientific™ Racks and Accessories for CO2 Resistant Shaker
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
Supplier:  Thermo Scientific™ 88881127
 View more versions of this product
Catalog No. 88881127
Only null left
Description
Description
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
  • Compatible with Thermo Scientific™ CO2 Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Specifications
Control Box
158 x 95 mm
6.2 x 3.7 x 1.0 in. (158 x 95 x 26 mm)
CO2 Resistant Shaker
Product Suggestions
Product Suggestions