Learn More
Thermo Scientific™ Racks and Accessories for CO2 Resistant Shaker
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
Supplier:  Thermo Scientific™ 88881119
 View more versions of this product
Catalog No. 88881119
Only null left
Description
Description
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
  • Compatible with Thermo Scientific™ CO2 Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Specifications
Fixed Tube Rack
40 x 16 mm dia.
CO2 Resistant Shaker
2
Product Suggestions
Product Suggestions