missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Racecadotril 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 89°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 385.48 |
| Percent Purity | ≥99.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Racecadotril |
Safety and Handling
| RTECSNumber | MB7606872 |
| TSCA | No |