Learn More
MilliporeSigma™ Rac1 Activation Magnetic Beads Pulldown Assay
Catalog No. 1710393
Change view
Click to view available options
Quantity:
30 Assays
Catalog No. Quantity
1710393 30 Assays
1 options
Catalog No. 1710393 Supplier MilliporeSigma™ Supplier No. 1710393
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
  • Rac1/Cdc42 Assay Reagent: 300ug of Pak-1 PBD in 150uL of glutathione magnetic beads, provided as a 50% beads slurry in PBS containing 50% glycerol for a final volume of 300uL.
  • Anti-Rac1, clone 23A8: One vial containing 250ug of purified IgG2akappa in 250uL of storage buffer (0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, containing 0.05% sodium azide).
  • Mg2+ Lysis/Wash Buffer, 5X.Two vials, each vial contains 18mL of 125mM, HEPES, pH 7.5, 750mM NaCl, 5% Igepal CA-630, 50mM MgCl2, 5mM EDTA and 10% glycerol
  • 100X GTPγS, 10mM, One vial containing 50 uL of 10mM GTPγS, 100X stock, in sterile water. Non-hydrolyzable analog of GTP. Sufficient to label 5mL of cell lysates
  • 100X GDP, 100mM. One vial containing 50uL of 100mM GDP, 100X stock, in sterile water. GDP, Guanosine 5'-Diphosphate, type I, for in vitro labeling of G-proteins in the inactive form. Sufficient to label 5ml of cell lysates.
Used to precipitate Rac1-GTP from cell lysates, detection by Rac1 specific monoclonal antibody
Can be used to precipitate Rac1-GTP from cell lysates, & detection by a Rac1 specific monoclonal antibody
Specifications
Quantity 30 Assays