missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit IgG Isotype Control

Catalog No. 026102
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
026102 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 026102 Supplier Invitrogen™ Supplier No. 026102
Only null left

Rabbit Polyclonal Isotype Control

Rabbit IgG is purified from pooled normal rabbit serum. Purity is verified by SDS-PAGE analysis. Concentration is 5.0 mg/mL based on an E (1%) = 14.0 at 280 nm.

Rabbit IgG refers to immunoglobulin G antibodies produced by rabbits in response to an antigen. These antibodies are commonly used in laboratory research as a detection tool for specific proteins, molecules, or cells. Rabbit IgG antibodies are highly specific and have a strong affinity for their target, making them ideal for use in various applications such as western blotting, immunohistochemistry, and flow cytometry.

Specifications

Antigen Rabbit IgG
Applications Control, Flow Cytometry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias Immunoglobulin G; ImmunoglobulinG; Not Applicable
Host Species Rabbit
Immunogen Gamma Immunoglobin
Purification Method Purified
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Isotype Control
Form Liquid
Isotype IgG
Show More Show Less