Recombinant Protein;1MG;Rabbit CD3E&CD3G Heterodimer Protein, His Tag&Flag Tag (CDG-RB52D5) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Thr 120 (CD3E) & Gln 23 - Asn 111 (CD3G) (Accession # Q9TUF9-1 (CD3E) & G1SDA3-1 (CD3G)).;Rabbit CD3E&CD3G Heterodimer Protein, His Tag&Flag Tag is produced by co-expression of CD3E and CD3G, has a calculated MW of 16.5 kDa (CD3E) and 15.0 kDa (CD3G). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3G is fused with a flag tag at the C-terminus. The reducing (R) heterodimer protein migrates as 18-20 kDa and 23 kDa due to glycosylation.