Learn More
Alkali Scientific R3005 Tube Roller with 5 Rolle
Catalog No. 503029614
Encompass
Gentle rolling motion Smooth 5 tilting notion Accepts tubes and bottles from 8mm to 200mm The Alkali ScientificR3005 Tuberoller are designed for the gentle thorough mixing of ra variety of containers including Vacutainers bottles test tubes centrifuge tubes and roller bottles Both models feature variable speed control (2-80 rpm) suitable for the broadest range of mixing applications As the roller mechanism rotates through each complete revolution the right side of each roller is gently elevated about 0 5 in (13mm) and returns to the horizontal starting position This additional gentle up/down motion provides a wave effect and guarantees that the entire sample is subjected to complete thorough mixing end to end This built-in tilt feature provides a level of homogeneous mixing uniformity that cannot be matched by any stationary roller apparatus From blood collection tubes to roller bottles Alkali TubeRollers (R3005) are the ideal choice for research clinical laboratories and doctor
Catalog No. 503029614 Supplier Alkali Scientific Supplier No. RS3005
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Gentle rolling motion Smooth 5 tilting notion Accepts tubes and bottles from 8mm to 200mm The Alkali ScientificR3005 Tuberoller are designed for the gentle thorough mixing of ra variety of containers including Vacutainers bottles test tubes centrifuge tubes and roller bottles Both models feature variable speed control (2-80 rpm) suitable for the broadest range of mixing applications As the roller mechanism rotates through each complete revolution the right side of each roller is gently elevated about 0 5 in (13mm) and returns to the horizontal starting position This additional gentle up/down motion provides a wave effect and guarantees that the entire sample is subjected to complete thorough mixing end to end This built-in tilt feature provides a level of homogeneous mixing uniformity that cannot be matched by any stationary roller apparatus From blood collection tubes to roller bottles Alkali TubeRollers (R3005) are the ideal choice for research clinical laboratories and doctor