missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-Tetrahydrofurfurylamine 98.0+%, TCI America™

Catalog No. T25515G
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25515G 5 g
T2551200MG 200 mg
T25511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T25515G Supplier TCI America Supplier No. T25515G

Chemical Identifiers

CAS 7202-43-9
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00192476
InChI Key YNOGYQAEJGADFJ-RXMQYKEDSA-N
Synonym r---tetrahydrofurfurylamine, r-tetrahydrofuran-2-yl methanamine, r-tetrahydrofurfurylamine, 2r-oxolan-2-ylmethanamine, 2r-tetrahydro-2-furanmethanamine, 2-furanmethanamine, tetrahydro-, 2r, 1-2r-tetrahydrofuran-2-yl methanamine, pubchem7001, 2r-tetrahydrofurfurylamine
PubChem CID 2734124
IUPAC Name [(2R)-oxolan-2-yl]methanamine
SMILES C1CC(OC1)CN

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 2943
Boiling Point 60°C
Formula Weight 101.15
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material (R)-(-)-Tetrahydrofurfurylamine