missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-Tetrahydrofuran-2-carboxylic Acid 98.0+%, TCI America™
$234.87 - $706.48
Chemical Identifiers
| CAS | 87392-05-0 |
|---|---|
| Molecular Formula | C5H8O3 |
| Molecular Weight (g/mol) | 116.116 |
| MDL Number | MFCD00211271 |
| InChI Key | UJJLJRQIPMGXEZ-SCSAIBSYSA-N |
| Synonym | r-+-2-tetrahydrofuroic acid, r-+-tetrahydro-2-furoic acid, r-tetrahydrofuran-2-carboxylic acid, r-+-tetrahydrofuran-2-carboxylic acid, r-2-tetrahydrofuroic acid, 2-furancarboxylic acid, tetrahydro-, 2r, 2r-oxolane-2-carboxylic acid, unii-7396hfq44j, r-tetrahydro-2-furoic acid, d-tetrahydro-furan-2-carboxylic acid |
| PubChem CID | 2734707 |
| IUPAC Name | (2R)-oxolane-2-carboxylic acid |
| SMILES | C1CC(OC1)C(=O)O |
Chemical Identifiers
| 87392-05-0 | |
| 116.116 | |
| UJJLJRQIPMGXEZ-SCSAIBSYSA-N | |
| 2734707 | |
| C1CC(OC1)C(=O)O |
| C5H8O3 | |
| MFCD00211271 | |
| r-+-2-tetrahydrofuroic acid, r-+-tetrahydro-2-furoic acid, r-tetrahydrofuran-2-carboxylic acid, r-+-tetrahydrofuran-2-carboxylic acid, r-2-tetrahydrofuroic acid, 2-furancarboxylic acid, tetrahydro-, 2r, 2r-oxolane-2-carboxylic acid, unii-7396hfq44j, r-tetrahydro-2-furoic acid, d-tetrahydro-furan-2-carboxylic acid | |
| (2R)-oxolane-2-carboxylic acid |
Specifications
| 87392-05-0 | |
| 129°C | |
| Clear Liquid at 20°C | |
| MFCD00211271 | |
| r-+-2-tetrahydrofuroic acid, r-+-tetrahydro-2-furoic acid, r-tetrahydrofuran-2-carboxylic acid, r-+-tetrahydrofuran-2-carboxylic acid, r-2-tetrahydrofuroic acid, 2-furancarboxylic acid, tetrahydro-, 2r, 2r-oxolane-2-carboxylic acid, unii-7396hfq44j, r-tetrahydro-2-furoic acid, d-tetrahydro-furan-2-carboxylic acid | |
| C1CC(OC1)C(=O)O | |
| 116.116 | |
| 116.12 | |
| (R)-(+)-Tetrahydrofuran-2-carboxylic Acid |
| Yellow | |
| 5 g | |
| C5H8O3 | |
| 3265 | |
| UJJLJRQIPMGXEZ-SCSAIBSYSA-N | |
| (2R)-oxolane-2-carboxylic acid | |
| 2734707 | |
| ≥98.0% (GC,T) |
Safety and Handling
TSCA : No