missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Tetrahydro-2-furoic acid, 98+%, ee 98%

Catalog No. p-7047541
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1941003 1 g
AAL1941006 5 g
AAL1941014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1941003 Supplier Thermo Scientific Chemicals Supplier No. L1941003

CAS: 87392-05-0 | C5H8O3 | 116.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87392-05-0
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00211271
InChI Key UJJLJRQIPMGXEZ-SCSAIBSYSA-N
Synonym r-+-2-tetrahydrofuroic acid, r-+-tetrahydro-2-furoic acid, r-tetrahydrofuran-2-carboxylic acid, r-+-tetrahydrofuran-2-carboxylic acid, r-2-tetrahydrofuroic acid, 2-furancarboxylic acid, tetrahydro-, 2r, 2r-oxolane-2-carboxylic acid, unii-7396hfq44j, r-tetrahydro-2-furoic acid, d-tetrahydro-furan-2-carboxylic acid
PubChem CID 2734707
IUPAC Name (2R)-oxolane-2-carboxylic acid
SMILES C1CC(OC1)C(=O)O

Specifications

Density 1.213
Quantity 1 g
Refractive Index 1.46
UN Number UN3265
Beilstein 4658739
Boiling Point 244°C
Formula Weight 116.12
Percent Purity ≥98%
Assay ee 98%
Chemical Name or Material (R)-(+)-Tetrahydro-2-furoic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only