missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-tert-Butylsulfinamide 98.0+%, TCI America™

Catalog No. B29075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29075G 5 g
B29071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29075G Supplier TCI America Supplier No. B29075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 196929-78-9
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.20
MDL Number MFCD05861479
InChI Key CESUXLKAADQNTB-UHFFFAOYNA-N
Synonym r-+-2-methyl-2-propanesulfinamide, r-2-methylpropane-2-sulfinamide, r-+-t-butylsulfinamide, r-+-tert-butanesulfinamide, r-+-tert-butylsulfinamide, r-2-methyl-2-propanesulfinamide, r-tert-butanesulfinamide, r-+-tert-butyl sulphinamide, r-tert-butylsulfinamide, r-+-tert-butyl sulfinamide
PubChem CID 10964479
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(N)=O

Specifications

Melting Point 106°C
Color White
Quantity 5 g
Formula Weight 121.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-tert-Butylsulfinamide