missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-tert-Butylsulfinamide, 98%

Catalog No. p-4228484
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391100010 1 g Glass bottle
AC391100050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC391100010 Supplier Thermo Scientific Chemicals Supplier No. 391100010
Only null left

CAS: 196929-78-9 | C4H11NOS | 121.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 196929-78-9
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.20
MDL Number MFCD05861479
InChI Key CESUXLKAADQNTB-UHFFFAOYNA-N
Synonym r-+-2-methyl-2-propanesulfinamide, r-2-methylpropane-2-sulfinamide, r-+-t-butylsulfinamide, r-+-tert-butanesulfinamide, r-+-tert-butylsulfinamide, r-2-methyl-2-propanesulfinamide, r-tert-butanesulfinamide, r-+-tert-butyl sulphinamide, r-tert-butylsulfinamide, r-+-tert-butyl sulfinamide
PubChem CID 10964479
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(N)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 99.0°C to 102.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CSONH2
Quantity 1 g
Merck Index 15,3607
Specific Rotation 3.0° to 5.0° (20°C, 589nm) (c=1, chloroform)
Formula Weight 121.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (R)-(+)-tert-Butylsulfinamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only