missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-SEGPHOS(regR) 99.0+%, TCI America™

Catalog No. S0930200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0930200MG 200 mg
S09301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0930200MG Supplier TCI America Supplier No. S0930200MG
Only null left

Chemical Identifiers

CAS 244261-66-3
Molecular Formula C38H28O4P2
Molecular Weight (g/mol) 610.59
MDL Number MFCD09753005
InChI Key RZZDRSHFIVOQAF-UHFFFAOYSA-N
Synonym (R)-(+)-5,5′C-Bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole, (R)-(+)-4,4′C-Bis(diphenylphosphino)-3,3′C-bi(1,2-methylenedioxybenzene)
PubChem CID 11017510
IUPAC Name {4-[5-(diphenylphosphanyl)-2H-1,3-benzodioxol-4-yl]-2H-1,3-benzodioxol-5-yl}diphenylphosphane
SMILES C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 172°C
Quantity 200 mg
Formula Weight 610.59
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-SEGPHOS(regR)