missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-sec-Butylamine 98.0+%, TCI America™

Catalog No. p-7119840
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2917100MG 100 mg
B29171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2917100MG Supplier TCI America Supplier No. B2917100MG

Chemical Identifiers

CAS 13250-12-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00064416
InChI Key BHRZNVHARXXAHW-SCSAIBSYSA-N
Synonym r---2-aminobutane, r---sec-butylamine, 2r-butan-2-amine, r-sec-butylamine, 2-butanamine, r, unii-29hc5icb6k, 2-butanamine, 2r, 29hc5icb6k, 2-butanamine, 2r-9ci, r-2-butanamine
PubChem CID 2724537
IUPAC Name (2R)-butan-2-amine
SMILES CCC(C)N

Specifications

Color Colorless
Boiling Point 63°C
Quantity 100 mg
UN Number 2733
Formula Weight 73.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(-)-sec-Butylamine
EINECSNumber (2)-0132
TSCA No