missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-S-Methyl-S-phenylsulfoximine, 97%

Catalog No. AAL08562MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL08562MD 250 mg
AAL0856203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL08562MD Supplier Thermo Scientific Chemicals Supplier No. L08562MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 60933-65-5 | C7H9NOS | 155.215 g/mol

(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 60933-65-5
Molecular Formula C7H9NOS
Molecular Weight (g/mol) 155.215
MDL Number MFCD00151460
InChI Key YFYIDTVGWCYSEO-JTQLQIEISA-N
Synonym s-+-s-methyl-s-phenylsulfoximine, s-+-s-methyl-s-phenylsulfoximine ee, s-imino methyl phenyl-??-sulfanone, r---s-methyl-s-phenylsulfoximine ee, imino-methyl-oxo-phenyl-$l^ 6, s-+-s-methyl-s-phenylsulphoximine, sulfoximine,s-methyl-s-phenyl-, s s
PubChem CID 25036288
IUPAC Name imino-methyl-oxo-phenyl-$l^{6}-sulfane
SMILES CS(=N)(=O)C1=CC=CC=C1

Specifications

Melting Point 29°C to 31°C
Quantity 250 mg
Beilstein 5245635
Sensitivity Hygroscopic
Optical Rotation −36° (c=1 in Acetone)
Formula Weight 155.22
Percent Purity 97%
Chemical Name or Material (R)-(-)-S-Methyl-S-phenylsulfoximine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only