missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-Ts-DENEB(regR), TCI America™

Catalog No. p-7137563
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3078200MG 200 mg
T30781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3078200MG Supplier TCI America Supplier No. T3078200MG
  • Offers superior activity and substrate scope compared to conventional RuCl(arene) N-sulfonylated diamine systems
    • Application: Selective asymmetric transfer hydrogenation of ketones
    • Performance: Reduces carbonyl groups selectively
    • Efficiency: Requires no high-pressure equipment by using formic acid or IPA as a hydrogen source

Chemical Identifiers

CAS 1333981-84-2
Molecular Formula C31H33ClN2O3RuS
Molecular Weight (g/mol) 650.196
MDL Number MFCD20922902
InChI Key INKUCOHLIHBSDN-ZAMYOOMVSA-M
Synonym Chloro[(R,R)-N-[2-[2-(4-methylbenzyloxy)ethyl]amino-1,2-diphenylethyl]-p-toluenesulfonamide]ruthenium(II), Chloro[(R,R)-N-[2-(4-methylbenzyloxy)ethyl]-N′C-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
PubChem CID 121235224
IUPAC Name chlororuthenium(1+);[(1R,2R)-2-[2-[(4-methylphenyl)methoxy]ethylamino]-1,2-diphenylethyl]-(4-methylphenyl)sulfonylazanide
SMILES CC1=CC=C(C=C1)COCCNC(C2=CC=CC=C2)C(C3=CC=CC=C3)[N-]S(=O)(=O)C4=CC=C(C=C4)C.Cl[Ru+]

Specifications

Color White-Yellow
Physical Form Crystalline Powder
Quantity 200 mg
UN Number 3077
Formula Weight 650.20
Concentration or Composition (by Analyte or Components) 12 to 17% Ruthenium
Packaging Glass Bottle
Product Line DENEB™
Sensitivity Air Sensitive
Appearance White to Brown Powder or Crystal
Decomposition Information Carbon Monoxide, Carbon Dioxide, Nitrogen Oxides, Hydrogen Chloride Gas, Metal Oxides
Chemical Name or Material (R,R)-Ts-DENEB(regR)
Show More Show Less