missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R,R,R)-Triglycidyl Isocyanurate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 106°C |
| Color | White |
| Quantity | 1 g |
| UN Number | 2811 |
| Formula Weight | 297.27 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (R,R,R)-Triglycidyl Isocyanurate |
Safety and Handling
| EINECSNumber | (5)-1052 |
| RTECSNumber | XZ1994900 |
| TSCA | No |