missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) Chloride 98.0+%, TCI America™
$298.08 - $928.43
Chemical Identifiers
| CAS | 138124-32-0 |
|---|---|
| Molecular Formula | C36H54ClMnN2O2- |
| Molecular Weight (g/mol) | 637.228 |
| MDL Number | MFCD02101664 |
| InChI Key | YRVXCOIDMFNGIJ-SEILFYAJSA-M |
| Synonym | 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene manganese iii chloride r,r-jacobsen cat. |
| PubChem CID | 131675872 |
| IUPAC Name | 2,4-ditert-butyl-6-[[[(1R,2R)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one;manganese;chloride |
| SMILES | CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Cl-].[Mn] |
Chemical Identifiers
| 138124-32-0 | |
| 637.228 | |
| YRVXCOIDMFNGIJ-SEILFYAJSA-M | |
| 131675872 | |
| CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Cl-].[Mn] |
| C36H54ClMnN2O2- | |
| MFCD02101664 | |
| 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene manganese iii chloride r,r-jacobsen cat. | |
| 2,4-ditert-butyl-6-[[[(1R,2R)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one;manganese;chloride |
Specifications
| 138124-32-0 | |
| C36H54ClMnN2O2- | |
| 1 g | |
| YRVXCOIDMFNGIJ-SEILFYAJSA-M | |
| 2,4-ditert-butyl-6-[[[(1R,2R)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one;manganese;chloride | |
| 131675872 | |
| ≥98.0% (HPLC) | |
| (R,R)-(-)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) Chlorid |
| 326°C | |
| MFCD02101664 | |
| 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene manganese iii chloride r,r-jacobsen cat. | |
| CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Cl-].[Mn] | |
| 637.228 | |
| 635.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No