missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II), 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 176763-62-5 |
|---|---|
| Molecular Formula | C36H52CoN2O2 |
| Molecular Weight (g/mol) | 603.76 |
| MDL Number | MFCD01631277 |
| InChI Key | XJWCTQVLJVAWMC-FAZMKCLSSA-N |
| Synonym | r,r-n,n'-bis 3,5-di-tert-butylsalicylidene-1,2-cyclohexanediaminocobalt ii |
| PubChem CID | 126963446 |
| IUPAC Name | cobalt;2,4-ditert-butyl-6-[[[(1R,2S)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one |
| SMILES | CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Co] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC329950010
|
Thermo Scientific Chemicals
329950010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC329950050
|
Thermo Scientific Chemicals
329950050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC329950250
|
Thermo Scientific Chemicals
329950250 |
25g | Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 176763-62-5 | |
| 603.76 | |
| XJWCTQVLJVAWMC-FAZMKCLSSA-N | |
| 126963446 | |
| CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Co] |
| C36H52CoN2O2 | |
| MFCD01631277 | |
| r,r-n,n'-bis 3,5-di-tert-butylsalicylidene-1,2-cyclohexanediaminocobalt ii | |
| cobalt;2,4-ditert-butyl-6-[[[(1R,2S)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one |
Specifications
| 176763-62-5 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| MFCD01631277 | |
| r,r-n,n'-bis 3,5-di-tert-butylsalicylidene-1,2-cyclohexanediaminocobalt ii | |
| XJWCTQVLJVAWMC-FAZMKCLSSA-N | |
| cobalt;2,4-ditert-butyl-6-[[[(1R,2S)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one | |
| 126963446 | |
| 98% |
| 97.5 | |
| 97.5% min. | |
| 98% | |
| C36H52CoN2O2 | |
| 1g | |
| Solubility in water: insoluble | |
| CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Co] | |
| 603.76 | |
| 603.76 | |
| (R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II) |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only