missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminocobalt(II), 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | >350°C |
| Color | Brown to Orange |
| Elemental Analysis | 97.5% min. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass Bottle |
| Quantity | 25 g |
| Solubility Information | Solubility in water: insoluble |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF |
RUO – Research Use Only