missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-2-Iodo-1,3-bis[1-(mesitylcarbamoyl)ethoxy]benzene 98.0+%, TCI America™

Catalog No. I0807200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0807200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0807200MG Supplier TCI America Supplier No. I0807200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1226896-38-3
Molecular Formula C30H35IN2O4
Molecular Weight (g/mol) 614.524
InChI Key ZVOKSLMZXDIXPR-DHIUTWEWSA-N
Synonym (R,R)-2-Iodo-1,3-bis[1-(2,4,6-trimethylphenylcarbamoyl)ethoxy]benzene
PubChem CID 46192097
IUPAC Name (2R)-2-[2-iodo-3-[(2R)-1-oxo-1-(2,4,6-trimethylanilino)propan-2-yl]oxyphenoxy]-N-(2,4,6-trimethylphenyl)propanamide
SMILES CC1=CC(=C(C(=C1)C)NC(=O)C(C)OC2=C(C(=CC=C2)OC(C)C(=O)NC3=C(C=C(C=C3C)C)C)I)C

Specifications

Color White
Quantity 200 mg
Formula Weight 614.52
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R,R)-2-Iodo-1,3-bis[1-(mesitylcarbamoyl)ethoxy]benzene
TSCA No
Recommended Storage Refrigerator