missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R,R)-2,6-Bis(4-phenyl-2-oxazolin-2-yl)pyridine 98.0+%, TCI America™
$119.30 - $326.21
Chemical Identifiers
| CAS | 128249-70-7 |
|---|---|
| Molecular Formula | C23H19N3O2 |
| Molecular Weight (g/mol) | 369.424 |
| MDL Number | MFCD01863585 |
| InChI Key | HLHBIMJNCKZZQO-SFTDATJTSA-N |
| Synonym | 2,6-bis 4r-4-phenyl-2-oxazolinyl pyridine, r,r-ph-pybox, 2,6-bis r-4-phenyl-4,5-dihydrooxazol-2-yl pyridine, +-2,6-bis 4r-4-phenyl-2-oxazolin-2-yl pyridine, ph-pybox, r,r, 2,6-bis 4r-phenyl-2-oxazolin-2-yl pyridine, r,r-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine, unii-555pn397nx, r,r-2,2'-2,6-pyridinediyl bis 4-phenyl-2-oxazoline, 2,6-bis 4r-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine |
| PubChem CID | 2734679 |
| IUPAC Name | (4R)-4-phenyl-2-[6-[(4R)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole |
| SMILES | C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B2219250MG
|
TCI America
B2219250MG |
250 mg |
Each for $119.30
|
|
|||||
|
B22191G
|
TCI America
B22191G |
1 g |
Each for $326.21
|
|
|||||
Chemical Identifiers
| 128249-70-7 | |
| 369.424 | |
| HLHBIMJNCKZZQO-SFTDATJTSA-N | |
| 2734679 | |
| C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5 |
| C23H19N3O2 | |
| MFCD01863585 | |
| 2,6-bis 4r-4-phenyl-2-oxazolinyl pyridine, r,r-ph-pybox, 2,6-bis r-4-phenyl-4,5-dihydrooxazol-2-yl pyridine, +-2,6-bis 4r-4-phenyl-2-oxazolin-2-yl pyridine, ph-pybox, r,r, 2,6-bis 4r-phenyl-2-oxazolin-2-yl pyridine, r,r-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine, unii-555pn397nx, r,r-2,2'-2,6-pyridinediyl bis 4-phenyl-2-oxazoline, 2,6-bis 4r-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine | |
| (4R)-4-phenyl-2-[6-[(4R)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole |
Specifications
| 128249-70-7 | |
| White-Yellow | |
| MFCD01863585 | |
| 2,6-bis 4r-4-phenyl-2-oxazolinyl pyridine, r,r-ph-pybox, 2,6-bis r-4-phenyl-4,5-dihydrooxazol-2-yl pyridine, +-2,6-bis 4r-4-phenyl-2-oxazolin-2-yl pyridine, ph-pybox, r,r, 2,6-bis 4r-phenyl-2-oxazolin-2-yl pyridine, r,r-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine, unii-555pn397nx, r,r-2,2'-2,6-pyridinediyl bis 4-phenyl-2-oxazoline, 2,6-bis 4r-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine | |
| C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5 | |
| 369.424 | |
| 369.42 | |
| Crystalline Powder |
| 173°C | |
| C23H19N3O2 | |
| 250 mg | |
| HLHBIMJNCKZZQO-SFTDATJTSA-N | |
| (4R)-4-phenyl-2-[6-[(4R)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole | |
| 2734679 | |
| ≥98.0% (HPLC,T) | |
| (R,R)-2,6-Bis(4-phenyl-2-oxazolin-2-yl)pyridine |
Safety and Handling
TSCA : No