missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 154°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Synonym | (R,R)-2,2′-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline) |
| Formula Weight | 301.39 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine |
Safety and Handling
| TSCA | No |