missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine 98.0+%, TCI America™

Catalog No. B2217250MG
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2217250MG 250 mg
B22171G 1 g
B22175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B2217250MG Supplier TCI America Supplier No. B2217250MG
Only null left

Chemical Identifiers

CAS 131864-67-0
Molecular Formula C17H23N3O2
Molecular Weight (g/mol) 301.39
MDL Number MFCD01321334
InChI Key CSGQGLBCAHGJDR-GJZGRUSLSA-N
PubChem CID 688213
IUPAC Name (4R)-4-propan-2-yl-2-[6-[(4R)-4-propan-2-yl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)C1COC(=N1)C2=NC(=CC=C2)C3=NC(CO3)C(C)C

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 250 mg
Synonym (R,R)-2,2′C-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline)
Formula Weight 301.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine