missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine, 97%

Catalog No. AAH27577MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27577MC 100 mg
AAH27577ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27577MC Supplier Thermo Scientific Chemicals Supplier No. H27577MC
Only null left

CAS: 90290-05-4 | C8H17NO2 | 159.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90290-05-4
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.229
MDL Number MFCD00210015
InChI Key HDXYTIYVVNJFLU-HTQZYQBOSA-N
Synonym 2r,5r-2,5-bis methoxymethyl pyrrolidine, r,r---2,5-bis methoxymethyl pyrrolidine, pyrrolidine, 2,5-bis methoxymethyl-, 2r,5r, 2r,5r-5-methoxymethyl pyrrolidin-2-yl methoxymethane, r,r---2,5-bis methoxymethyl-pyrrolidine, r,r-2,5-bis methoxymethyl pyrrolidine, 2beta,5alpha-bis methoxymethyl pyrrolidine
PubChem CID 2724974
IUPAC Name (2R,5R)-2,5-bis(methoxymethyl)pyrrolidine
SMILES COCC1CCC(N1)COC

Specifications

Boiling Point 106°C to 108°C (18 mmHg)
Refractive Index 1.449
Quantity 100 mg
Optical Rotation −8° (c=3 in Ethanol)
Formula Weight 159.23
Percent Purity 97%
Chemical Name or Material (R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only