missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-(+)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) 98.0+%, TCI America™

Catalog No. I0795100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0795100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0795100MG Supplier TCI America Supplier No. I0795100MG
Only null left

Chemical Identifiers

CAS 131833-97-1
Molecular Formula C17H30N2O2
Molecular Weight (g/mol) 294.44
MDL Number MFCD07368371
InChI Key DPMGLJUMNRDNMX-UHFFFAOYNA-N
Synonym (R,R)-(+)-2,2′C-(Dimethylmethylene)bis(4-tert-butyl-2-oxazoline), (R,R)-(+)-2,2-Bis(4-tert-butyl-2-oxazolin-2-yl)propane
PubChem CID 688210
IUPAC Name 4-tert-butyl-2-[2-(4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)(C)C1COC(=N1)C(C)(C)C1=NC(CO1)C(C)(C)C

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 100 mg
Formula Weight 294.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R,R)-(+)-2,2′-Isopropylidenebis(4-tert-butyl-2-oxazoline)